Få mere at vide
Diisopropyl Fluorophosphate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Diisopropyl Fluorophosphate |
| CAS | 55-91-4 |
| Molekylær formel | C6 H14 F O3 P |
| Mængde | 2.5 g |
| Synonym | Phosphorofluoridic acid, bis(1-methylethyl) ester, Phosphorofluoridic acid, diisopropyl ester (8CI), DFF, DFP, DFP (pesticide), Difluorophate, Diflupyl, Diisopropoxyphosphoryl fluoride, Diisopropyl fluorophosphate, Diisopropyl phosphofluoridate, Diisopropyl phosphorofluoridate, Dyflos, Floropryl, Fluorodiisopropyl phosphate, Fluostigmine, Isoflurophate, Isopropyl fluophosphate ((C3H7O)2FPO), Isopropyl phosphorofluoridate ((C3H7O)2FPO), Neoglaucit, O,O-Diisopropyl phosphorofluoridate |
| InChI formel | InChI=1S/C6H14FO3P/c1-5(2)9-11(7,8)10-6(3)4/h5-6H,1-4H3 |
| SMIL | CC(C)OP(=O)(F)OC(C)C |
| IUPAC navn | 2-[fluoro(propan-2-yloxy)phosphoryl]oxypropane |
| Molekylvægt (g/mol) | 184.15 |
| Formel vægt | 184.0665 |