Få mere at vide
Diisononyl Phthalate-d4, TRC
Artikelnummer. 30373211
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30373211 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30373211 Leverandør Toronto Research Chemicals Leverandørnr. TRCD455227100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Phthalic Acid Di-isononyl Ester (3,4,5,6)-D4
CAS 1332965-90-8
Renhedsgrad noter HPLC
Molekylær formel C26 2H4 H38 O4
Mængde 100 mg
Synonym 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid, 1,2-bis(7-methyloctyl) ester, Diisononyl Phthalate-d4, Phthalic Acid Di-isononyl Ester (3,4,5,6)-D4
InChI formel InChI=1S/C26H42O4/c1-21(2)15-9-5-7-13-19-29-25(27)23-17-11-12-18-24(23)26(28)30-20-14-8-6-10-16-22(3)4/h11-12,17-18,21-22H,5-10,13-16,19-20H2,1-4H3/i11D,12D,17D,18D
SMIL [2H]c1c([2H])c([2H])c(C(=O)OCCCCCCC(C)C)c(C(=O)OCCCCCCC(C)C)c1[2H]
IUPAC navn bis(7-methyloctyl) 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate
Molekylvægt (g/mol) 422.63
Formel vægt 422.3334
Procent renhed >95