Få mere at vide
Diisodecyl Phthalate-d4, TRC
Artikelnummer. 30373963
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30373963 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30373963 Leverandør Toronto Research Chemicals Leverandørnr. TRCD455217100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Diisodecyl phthalate-d4
CAS 1346604-79-2
Renhedsgrad noter HPLC
Molekylær formel C28 2H4 H42 O4
Mængde 100 mg
Synonym 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid, 1,2-bis(8-methylnonyl) ester, Diisodecyl Phthalate-d4, Phthalic Acid Di-isodecyl Ester (3,4,5,6)-D4
InChI formel InChI=1S/C28H46O4/c1-23(2)17-11-7-5-9-15-21-31-27(29)25-19-13-14-20-26(25)28(30)32-22-16-10-6-8-12-18-24(3)4/h13-14,19-20,23-24H,5-12,15-18,21-22H2,1-4H3/i13D,14D,19D,20D
SMIL [2H]c1c([2H])c([2H])c(C(=O)OCCCCCCCC(C)C)c(C(=O)OCCCCCCCC(C)C)c1[2H]
IUPAC navn bis(8-methylnonyl) 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate
Molekylvægt (g/mol) 450.69
Formel vægt 450.3647
Procent renhed >95