missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diisobutylaluminum hydride, 25% w/w in hexane, Thermo Scientific™
Click to view available options
Quantity:
50 mL
250 mL
1 L
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Color | Colorless |
Density | 0.701g/mL |
Linear Formula | [(CH3)2CHCH2]2AlH |
Quantity | 50 mL |
UN Number | UN3399 |
Sensitivity | Air & Moisture Sensitive |
Formula Weight | 142.22 |
Physical Form | Liquid |
Chemical Name or Material | Diisobutylaluminum hydride |
Safety and Handling
Product Identifier
- Diisobutylaluminum hydride, 25% w/w in hexane
- Danger
- Flammable liquid Category 1
- Substance which\, in contact with water\, emits flammable gases Category 1
- Skin corrosion/irritation Category 1 B
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
- Aspiration toxicity Category 1
- LONG-TERM AQUATIC HAZARD Chronic 2
- H224-Extremely flammable liquid and vapour.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H314-Causes severe skin burns and eye damage.
- H361-Suspected of damaging fertility or the unborn child.
- H336-May cause drowsiness or dizziness.
- H373-May cause damage to organs.
- H304-May be fatal if swallowed and enters airways.
- H411-Toxic to aquatic life with long lasting effects.
- EUH014-Reacts violently with water.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'