Få mere at vide
Diisobutyl Phthalate-d4, Diisodecyl Phthalate-d4, and Diisononyl Phthalate-d4 Mixture (0.1 mg/mL in Acetonitrile), TRC
Artikelnummer. 30466348
Skift visning
Mængde:
1 mL
Artikelnummer. Mængde
30466348 1 mL
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30466348 Leverandør Toronto Research Chemicals Leverandørnr. TRCKIT79221X1ML
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Diisodecyl phthalate-d4∣Phthalic Acid Di-isononyl Ester (3,4,5,6)-D4∣Phthalic acid, bis-iso-butyl ester D4
CAS 1346604-79-2, 1332965-90-8, 358730-88-8
Opløsningsmiddel eller Matrix Acetonitrile
Mængde 1 mL
Koncentration eller sammensætning (efter analyt eller komponenter) Diisodecyl phthalate-d4 0.1mg/mL∣Phthalic Acid Di-isononyl Ester (3,4,5,6)-D4 0.1mg/mL∣Phthalic acid, bis-iso-butyl ester D4 0.1mg/mL