Få mere at vide
Dihydrostreptomycin 1000 μg/mL in Water, Dr. Ehrenstorfer
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Dihydrostreptomycin |
| CAS | 128-46-1 |
| Analyt- eller komponentnavne | Dihydrostreptomycin |
| Forsendelsestilstand | +20°C |
| Molekylær formel | C21 H41 N7 O12 |
| Koncentration | 1000 μg/mL |
| Certificeringer/compliance | ISO 17034 |
| Synonym | D-Streptamine, O-2-deoxy-2-(methylamino)-α-L-glucopyranosyl-(1→2)-O-5-deoxy-3-C-(hydroxymethyl)-α-L-lyxofuranosyl-(1→4)-N1,N3-bis(aminoiminomethyl)-, D-Streptamine, O-2-deoxy-2-(methylamino)-α-L-glucopyranosyl-(1→2)-O-5-deoxy-3-C-(hydroxymethyl)-α-L-lyxofuranosyl-(1→4)-N,N'-bis(aminoiminomethyl)- (9CI), Streptomycin, deoxydihydrohydroxy- (6CI,7CI), Streptomycin, dihydro- (8CI), O-2-Deoxy-2-(methylamino)-α-L-glucopyranosyl-(1→2)-O-5-deoxy-3-C-(hydroxymethyl)-α-L-lyxofuranosyl-(1→4)-N1,N3-bis(aminoiminomethyl)-D-streptamine, 2,4-Diguanidino-3,5,6-trihydroxycyclohexyl-5-deoxy-2-O-(2-deoxy-2-methylamino-α-L-glucopyranosyl)-3-hydroxymethyl-β-L-lyxo-pentanofuranoside, Abiocine, DHSM, DST, Dihydrostreptomycin, Vibriomycin |
| InChI formel | InChI=1S/C21H41N7O12/c1-5-21(36,4-30)16(40-17-9(26-2)13(34)10(31)6(3-29)38-17)18(37-5)39-15-8(28-20(24)25)11(32)7(27-19(22)23)12(33)14(15)35/h5-18,26,29-36H,3-4H2,1-2H3,(H4,22,23,27)(H4,24,25,28)/t5-,6-,7+,8-,9-,10-,11+,12-,13-,14+,15+,16-,17-,18-,21+/m0/s1 |
| SMIL | CN[C@H]1[C@H](O)[C@@H](O)[C@H](CO)O[C@H]1O[C@H]2[C@H](O[C@H]3[C@H](O)[C@@H](O)[C@H](NC(=N)N)[C@@H](O)[C@@H]3NC(=N)N)O[C@@H](C)[C@]2(O)CO |
| Vis mere |