Få mere at vide
Dihydrolysergic Acid Methyl Ester, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Dihydrolysergic Acid Methyl Ester |
| CAS | 35470-53-2 |
| Molekylær formel | C17H20N2O2 |
| Mængde | 5 mg |
| Synonym | Methyl 6-Methylergoline-8beta-carboxylate, Methyl 9,10-Dihydro-D-lysergate, Methyl 9,10-Dihydrolysergate, Methyl D-6-Methylergoline-8beta-carboxylate, Methyl Dihydrolysergate, 9,10-Dihydrolysergic Acid Methyl EsterD-Dihydrolysergic Acid Methyl Ester |
| InChI formel | InChI=1S/C17H20N2O2/c1-19-9-11(17(20)21-2)6-13-12-4-3-5-14-16(12)10(8-18-14)7-15(13)19/h3-5,8,11,13,15,18H,6-7,9H2,1-2H3/t11-,13-,15-/m1/s1 |
| SMIL | COC(=O)[C@@H]1C[C@H]2[C@@H](Cc3c[nH]c4cccc2c34)N(C)C1 |
| IUPAC navn | methyl (6aR,9R,10aR)-7-methyl-6,6a,8,9,10,10a-hexahydro-4H-indolo[4,3-fg]quinoline-9-carboxylate |
| Molekylvægt (g/mol) | 284.35 |
| Formel vægt | 284.1525 |