missing translation for 'onlineSavingsMsg'
Learn More

Dihydrogen hexachloroosmate(IV) hydrate, Premion™, 99.95% (metals basis), Os 37% min, Thermo Scientific Chemicals

Catalog Number p-7048952
Click to view available options
Quantity:
0.5 g
1 g
5 g
This item is not returnable. View return policy
27057-71-2
H2Cl6Os
404.93
MFCD00011329
GCRLVKBHFZOVLQ-UHFFFAOYSA-H
Hydrogen hexachloroosmate(IV); Chloroosmic acid
hexachloroosmium
Cl[Os](Cl)(Cl)(Cl)(Cl)Cl
This item is not returnable. View return policy
27057-71-2
H2Cl6Os
404.93
MFCD00011329
GCRLVKBHFZOVLQ-UHFFFAOYSA-H
Hydrogen hexachloroosmate(IV); Chloroosmic acid
hexachloroosmium
Cl[Os](Cl)(Cl)(Cl)(Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point (decomposition)
Color Black
Linear Formula H2OsCl6·xH2O
Quantity 0.5 g
UN Number UN3260
Sensitivity Hygroscopic
Formula Weight 404.93 (Anhydrous)
Percent Purity 99.95%
Grade Premion™
Concentration or Composition (by Analyte or Components) Os 37% min.
Physical Form Crystalline
Assay (metals basis)
Chemical Name or Material Dihydrogen hexachloroosmate(IV) hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'