Få mere at vide
Dihydroergotamine (+)-Tartrate Salt (2:1), TRC
Artikelnummer. 17984205
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17984205 50 mg
17970815 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17984205 Leverandør Toronto Research Chemicals Leverandørnr. TRCD45065350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 5989-77-5
Molekylær formel 2 C33 H37 N5 O5 . C4 H6 O6
Molekylvægt (g/mol) 1317.44
Synonym Dihydroergotamine tartrate, Bis[(6aR,9R,10aR)-N-[(2R,5S,10aS,10bS)-5-benzyl-10b-hydroxy-2-methyl-3,6-dioxooctahydro-8H-oxazolo[3,2-a]pyrrolo[2,1-c]pyrazin-2-yl]-7-methyl-4,6,6a,7,8,9,10,10a-octahydroindolo[4,3-fg]quinoline-9-carboxamide] (2R,3R)-2,3-dihydroxybutanedioate
IUPAC navn Bis[(6aR,9R,10aR)-N-[(2R,5S,10aS,10bS)-5-benzyl-10b-hydroxy-2-methyl-3,6-dioxooctahydro-8H-oxazolo[3,2-a]pyrrolo[2,1-c]pyrazin-2-yl]-7-methyl-4,6,6a,7,8,9,10,10a-octahydroindolo[4,3-fg]quinoline-9-carboxamide] (2R,3R)-2,3-dihydroxybutanedioate
SMIL CN1C[C@@H](C[C@H]2[C@H]1Cc3c[nH]c4cccc2c34)C(=O)N[C@]5(C)O[C@@]6(O)[C@@H]7CCCN7C(=O)[C@H](Cc8ccccc8)N6C5=O.CN9C[C@@H](C[C@H]%10[C@H]9Cc%11c[nH]c%12cccc%10c%11%12)C(=O)N[C@]%13(C)O[C@@]%14(O)[C@@H]%15CCCN%15C(=O)[C@H](Cc%16ccccc%16)N%14C%13=O.O[C@H]([C@@H](O)C(=O)O)C(=O)O
Tekniske data
Kemisk navn eller materiale Dihydroergotamine tartrate
Mængde 50 mg
InChI formel InChI=1S/2C33H37N5O5.C4H6O6/c2*1-32(35-29(39)21-15-23-22-10-6-11-24-28(22)20(17-34-24)16-25(23)36(2)18-21)31(41)38-26(14-19-8-4-3-5-9-19)30(40)37-13-7-12-27(37)33(38,42)43-32;5-1(3(7)8)2(6)4(9)10/h2*3-6,8-11,17,21,23,25-27,34,42H,7,12-16,18H2,1-2H3,(H,35,39);1-2,5-6H,(H,7,8)(H,9,10)/t2*21-,23-,25-,26+,27+,32-,33+;1-,2-/m111/s1
Formel vægt 1316.5754