Få mere at vide
Dihydroergotamin Mesylat, TRC
Artikelnummer. 30378796
Skift visning
Mængde:
5 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30378796 100 mg
30384033 5 mg
30377314 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30378796 Leverandør Toronto Research Chemicals Leverandørnr. TRCD449080100MG
 Begrænset vare
Forventet forsendelse: 09-12-2026
Højrenheds organiske molekyler og analytiske standarder, strategisk leveret verden over for at muliggøre innovation og kommerciel succes.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 6190-39-2
Molekylær formel C33 H37 N5 O5 . C H4 O3 S
Molekylvægt (g/mol) 679.78
Synonym Dihydroergotamine mesilate, (6aR,9R,10aR)-N-[(2R,5S,10aS,10bS)-5-Benzyl-10b-hydroxy-2-methyl-3,6-dioxooctahydro-8H-oxazolo[3,2-a]pyrrolo[2,1-c]pyrazin-2-yl]-7-methyl-4,6,6a,7,8,9,10,10a- octahydroindolo[4,3-fg]quinoline-9-carboxamide methanesulfonate, Ergotaman-3',6',18-trione, 9,10-dihydro-12'-hydroxy-2'-methyl-5'-(phenylmethyl)-, (5'α,10α)-, methanesulfonate (1:1), Ergotaman-3',6',18-trione, 9,10-dihydro-12'-hydroxy-2'-methyl-5'-(phenylmethyl)-, (5'α,10α)-, monomethanesulfonate (salt) (9CI), Ergotamine, 9,10-dihydro-, monomethanesulfonate (salt) (6CI,7CI,8CI), (+)-Dihydroergotamine mesylate, 9,10-Dihydroergotamine mesylate, Agit, Angionorm, DETMS, DHE 45, Dergotamine, Diergo, Dihydergot, Dihydroergotamine mesylate, Dihydroergotamine methanesulfonate, Dihydroergotamine methanesulphonate, Dihydroergotamine monomethanesulfonate, Dihytamine, Dirgotarl, Endophleban, Ergomimet, Ergont, Ergotonin, Ikaran, Migranal, Morena, Orstanorm, Seglor, Tonopres, Verladyn
IUPAC navn (6aR,9R,10aR)-N-[(2R,5S,10aS,10bS)-5-Benzyl-10b-hydroxy-2-methyl-3,6-dioxooctahydro-8H-oxazolo[3,2-a]pyrrolo[2,1-c]pyrazin-2-yl]-7-methyl-4,6,6a,7,8,9,10,10a-octahydroindolo[4,3-fg]quinolin-9-carboxamid metanesulfonat
SMIL CN1C[C@@H](C[C@H]2[C@H]1Cc3c[nH]c4cccc2c34)C(=O)N[C@]5(C)O[C@@]6(O)[C@@H]7CCCN7C(=O)[C@H](Cc8ccccc8)N6C5=O.CS(=O)(=O)O
Tekniske data
Kemisk navn eller materiale Dihydroergotamine mesilate
Mængde 100 mg
InChI formel InChI=1S/C33H37N5O5.CH4O3S/c1-32(35-29(39)21-15-23-22-10-6-11-24-28(22)20(17-34-24)16-25(23)36(2)18-21)31(41)38-26(14-19-8-4-3-5-9-19)30(40)37-13-7-12-27(37)33(38,42)43-32;1-5(2,3)4/h3-6,8-11,17,21,23,25-27,34,42H,7,12-16,18H2,1-2H3,(H,35,39);1H3,(H,2,3,4)/t21-,23-,25-,26+,27+,32-,33+;/m1./s1
Formel vægt 679.2676