Få mere at vide
Dihydro Uracil-d4, TRC
Artikelnummer. 30375282
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30375282 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30375282 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4499931MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Dihydro Uracil-d4
CAS 334473-41-5
Molekylær formel C4 D4 H2 N2 O2
Mængde 1 mg
Synonym 2,4(1H,3H)-Pyrimidinedione-5,6-d2, dihydro-d2- (9CI), Dihydro-d2-2,4(1H,3H)-pyrimidinedione-5,6-d2, Hydrouracil-d4, 5,6-Dihydro-2,4(1H,3H)-pyrimidinedione-5,5,6,6-d4, 5,6-Dihydro-2,4-dihydroxypyrimidine-5,5,6,6-d4, 5,6-Dihydrouracil-d4, 5,6-Dihydrouracil-5,5,6,6-D4, Dihydrouracil-d4
InChI formel InChI=1S/C4H6N2O2/c7-3-1-2-5-4(8)6-3/h1-2H2,(H2,5,6,7,8)/i1D2,2D2
SMIL [2H]C1([2H])NC(=O)NC(=O)C1([2H])[2H]
IUPAC navn 5,5,6,6-tetradeuterio-1,3-diazinane-2,4-dione
Molekylvægt (g/mol) 118.127
Formel vægt 118.068