Få mere at vide
Dihydro Nicotyrine-d3, TRC
Artikelnummer. 30379220
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30379220 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30379220 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4491741MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Dihydro Nicotyrine-d3
CAS 1246818-62-1
Molekylær formel C15 H19 N O8
Mængde 1 mg
Synonym 3-(4,5-Dihydro-1-[(methyl-d3)-1H-pyrrol-2-yl)]pyridine, 3-[4,5-Dihydro-1-(methyl-d3)pyrrol-2-yl]pyridine, N-Methylmyosmine-d3
InChI formel InChI=1S/C15H19NO8/c1-7(17)16-8-3-5-9(6-4-8)23-15-12(20)10(18)11(19)13(24-15)14(21)22-2/h3-6,10-13,15,18-20H,1-2H3,(H,16,17)/t10?,11-,12?,13?,15-/m1/s1
SMIL COC(=O)C1O[C@@H](Oc2ccc(NC(=O)C)cc2)C(O)C(O)[C@H]1O
IUPAC navn methyl (3R,6S)-6-(4-acetamidophenoxy)-3,4,5-trihydroxyoxane-2-carboxylate
Molekylvægt (g/mol) 341.313
Formel vægt 341.111