Få mere at vide
Dihydro Folic Acid, TRC
Artikelnummer. 30003992
Skift visning
Mængde:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30003992 25 mg
17927834 10 mg
17997354 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30003992 Leverandør Toronto Research Chemicals Leverandørnr. TRCD44970025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 4033-27-6
Molekylær formel C19 H21 N7 O6
Molekylvægt (g/mol) 443.41
Synonym L-Glutamic acid, N-[4-[[(2-amino-3,4,7,8-tetrahydro-4-oxo-6-pteridinyl)methyl]amino]benzoyl]-, Glutamic acid, N-[p-[[(2-amino-7,8-dihydro-4-hydroxy-6-pteridinyl)methyl]amino]benzoyl]-, L- (6CI,7CI,8CI), L-Glutamic acid, N-[4-[[(2-amino-1,4,7,8-tetrahydro-4-oxo-6-pteridinyl)methyl]amino]benzoyl]- (9CI), 7,8-Dihydro-L-folic acid, 7,8-Dihydrofolic acid, Dihydrofolic acid, NSC 165989, (2S)-2-[4-[[(2-Amino-4-oxo-1,4,7,8-tetrahydropteridin-6-yl)methyl]amino]benzamido]pentanedioic acid, (2S)-2-[[4-[[(2-Amino-4-oxo-1,4,7,8-tetrahydropteridin-6-yl)methyl]amino]benzoyl]amino]pentanedioic acid
IUPAC navn (2S)-2-[[4-[(2-amino-4-oxo-7,8-dihydro-1H-pteridin-6-yl)methylamino]benzoyl]amino]pentanedioic acid
SMIL NC1=NC(=O)C2=C(NCC(=N2)CNc3ccc(cc3)C(=O)N[C@@H](CCC(=O)O)C(=O)O)N1
Tekniske data
Kemisk navn eller materiale Dihydro Folic Acid
InChI formel InChI=1S/C19H21N7O6/c20-19-25-15-14(17(30)26-19)23-11(8-22-15)7-21-10-3-1-9(2-4-10)16(29)24-12(18(31)32)5-6-13(27)28/h1-4,12,21H,5-8H2,(H,24,29)(H,27,28)(H,31,32)(H4,20,22,25,26,30)/t12-/m0/s1
Mængde 25 mg
Formel vægt 443.1553
Forsendelsestilstand Dry Ice Dry Pack