missing translation for 'onlineSavingsMsg'
Få mere at vide

Dihexadecyl(dimethyl)azanium Bromide, TRC

Artikelnummer. 30340442
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
2.5 g
Pakningsstørrelse:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30340442 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30340442 Leverandør Toronto Research Chemicals Leverandørnr. TRCB4264452.5G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Dihexadecyl(dimethyl)azanium Bromide
CAS 70755-47-4
Molekylær formel C34H72BrN
Mængde 2.5 g
Synonym 1-Hexadecanaminium, N-hexadecyl-N,N-dimethyl-, bromide (9CI), Bis(hexadecyl) dimethyl ammonium bromide, Di-n-hexadecyldimethylammonium bromide, Dicetyldimethylammonium bromide, Dicetyldimonium bromide, Dihexadecyldimethylammonium bromide, Dimethyldihexadecylammonium bromide, Dimethyldipalmitylammonium bromide, Dipalmityldimethylammonium bromide, Konstanta A, Dimethyldihexadecylammonium Bromide, N-hexadecyl-N,N-dimethyl-1-Hexadecanaminium bromide, Bis(hexadecyl) dimethyl ammonium bromide, Di-n-hexadecyldimethylammonium bromide, Dicetyldimethylammonium bromide, Dicetyldimonium bromide, Dihexadecyldimethylammonium bromide, Dimethyldihexadecylammonium bromide, Dimethyldipalmitylammonium bromide, Dipalmityldimethylammonium bromide, Konstanta A
InChI formel InChI=1S/C34H72N.BrH/c1-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35(3,4)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-2;/h5-34H2,1-4H3;1H/q+1;/p-1
SMIL [Br-].CCCCCCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCCCCCC
IUPAC navn dihexadecyl(dimethyl)azanium;bromide
Molekylvægt (g/mol) 574.85
Formel vægt 573.4848
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.