missing translation for 'onlineSavingsMsg'
Få mere at vide

Dihexadecyl(dimethyl)azanium Bromide, TRC

Artikelnummer. 30332797
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30332797 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30332797 Leverandør Toronto Research Chemicals Leverandørnr. TRCB426445250MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Dihexadecyl(dimethyl)azanium Bromide
CAS 70755-47-4
Molekylær formel C34H72BrN
Mængde 250 mg
Synonym 1-Hexadecanaminium, N-hexadecyl-N,N-dimethyl-, bromide (9CI), Bis(hexadecyl) dimethyl ammonium bromide, Di-n-hexadecyldimethylammonium bromide, Dicetyldimethylammonium bromide, Dicetyldimonium bromide, Dihexadecyldimethylammonium bromide, Dimethyldihexadecylammonium bromide, Dimethyldipalmitylammonium bromide, Dipalmityldimethylammonium bromide, Konstanta A, Dimethyldihexadecylammonium Bromide, N-hexadecyl-N,N-dimethyl-1-Hexadecanaminium bromide, Bis(hexadecyl) dimethyl ammonium bromide, Di-n-hexadecyldimethylammonium bromide, Dicetyldimethylammonium bromide, Dicetyldimonium bromide, Dihexadecyldimethylammonium bromide, Dimethyldihexadecylammonium bromide, Dimethyldipalmitylammonium bromide, Dipalmityldimethylammonium bromide, Konstanta A
InChI formel InChI=1S/C34H72N.BrH/c1-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35(3,4)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-2;/h5-34H2,1-4H3;1H/q+1;/p-1
SMIL [Br-].CCCCCCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCCCCCC
IUPAC navn dihexadecyl(dimethyl)azanium;bromide
Molekylvægt (g/mol) 574.85
Formel vægt 573.4848
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.