missing translation for 'onlineSavingsMsg'
Få mere at vide

Digoxigenin, MedChemExpress

Artikelnummer. 16468648
Encompass_Preferred
Skift visning
Click to view available options
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
16468648 10 mg
16434527 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16468648 Supplier Medchem Express Supplier No. HYB1025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Digoxigenin (DIG) is a steroid. DIG is used for situ hybridization as a labeling molecule probe due to long shelf life and fast detection and high sensitivity of DIG-labeled riboprobes.

Digoxigenin (DIG) is a steroid. DIG is used for situ hybridization as a labeling molecule probe due to long shelf life and fast detection and high sensitivity of DIG-labeled riboprobes.

Specifications

Kemisk navn eller materiale Digoxigenin
Fysisk form Solid
CAS 1672-46-4
Farve Off-White
Mængde 10 mg
Renhedsgrad noter Research
Molekylær formel C23H34O5
Til brug med (applikation) COVID-19-immunoregulation
Synonym Lanadigenin
Opløselighedsinformation DMSO : 125 mg/mL (320.09 mM; Need ultrasonic)
SMIL O=C1OCC([C@H]2CC[C@]3(O)[C@]4([H])CC[C@]5([H])C[C@@H](O)CC[C@]5(C)[C@@]4([H])C[C@@H](O)[C@]23C)=C1
Molekylvægt (g/mol) 390.51
Formel vægt 390.51
Procent renhed 98.0%
Grad Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.