Få mere at vide
Diglyme-d6, TRC
Artikelnummer. 30375211
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30375211 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30375211 Leverandør Toronto Research Chemicals Leverandørnr. TRCD44630210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Diglyme-d6
CAS 1189942-73-1
Molekylær formel C6H8D6O3
Mængde 10 mg
Synonym 1,1'-Oxybis[2-methoxyethane]-d6, Bis(2-methoxyethyl)-d6 Ether, 1,5-(Dimethoxy-d6)-3-oxapentane, 1-(2-Methoxyethoxy)-2-methoxyethane-d6, 2,5,8-Trioxanonane-d6, Diethylene Glycol Dimethyl-d6 Ether, Dimethyl-d6 Carbitol, Dimethyl-d6 Digol, Glyme 2-d6, Glyme 3-d6, Hisolve MDM-d6, NSC 59726-d6
InChI formel InChI=1S/C6H14O3/c1-7-3-5-9-6-4-8-2/h3-6H2,1-2H3/i1D3,2D3
SMIL [2H]C([2H])([2H])OCCOCCOC([2H])([2H])[2H]
IUPAC navn 1-(methoxy-d3)-2-(2-(methoxy-d3)ethoxy)ethane
Molekylvægt (g/mol) 140.21
Formel vægt 140.13