Få mere at vide
Digitoxin 3' ' '-Acetate, TRC
Artikelnummer. 30134826
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134826 1 mg
30004032 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30134826 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4453851MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1111-39-3
Molekylær formel C43 H66 O14
Molekylvægt (g/mol) 806.98
Synonym Card-20(22)-enolide, 3-[(O-3-O-acetyl-2,6-dideoxy-β-D-ribo-hexopyranosyl-(1→4)-O-2,6-dideoxy-β-D-ribo-hexopyranosyl-(1→4)-2,6-dideoxy-β-D-ribo-hexopyranosyl)oxy]-14-hydroxy-, (3β,5β)-, Digitoxin, 3' ' '-acetate (8CI), Digitoxin, α-acetyl- (6CI), (3β,5β)-3-[(O-3-O-Acetyl-2,6-dideoxy-β-D-ribo-hexopyranosyl-(1→4)-O-2,6-dideoxy-β-D-ribo-hexopyranosyl-(1→4)-2,6-dideoxy-β-D-ribo-hexopyranosyl)oxy]-14-hydroxycard-20(22)-enolide, Acetyldigitoxin, Acylanid, Adicin, Desglucolanatoside A, α-Acetyldigitoxin, α-Monoacetyldigitoxin
IUPAC navn [(2R,3R,4S,6S)-3-hydroxy-6-[(2R,3S,4S,6S)-4-hydroxy-6-[(2R,3S,4S,6R)-4-hydroxy-6-[[(3S,5R,8R,9S,10S,13R,14S,17R)-14-hydroxy-10,13-dimethyl-17-(5-oxo-2H-furan-3-yl)-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-3-yl]oxy]-2-methyloxan-3-yl]oxy-2-methyloxan-3-yl]oxy-2-methyloxan-4-yl] acetate
SMIL C[C@H]1O[C@H](C[C@H](OC(=O)C)[C@@H]1O)O[C@H]2[C@@H](O)C[C@H](O[C@H]3[C@@H](O)C[C@H](O[C@H]4CC[C@@]5(C)[C@H](CC[C@@H]6[C@@H]5CC[C@]7(C)[C@H](CC[C@]67O)C8=CC(=O)OC8)C4)O[C@@H]3C)O[C@@H]2C
Tekniske data
Kemisk navn eller materiale alpha-Acetyldigitoxin
Mængde 1 mg
InChI formel InChI=1S/C43H66O14/c1-21-38(48)33(54-24(4)44)19-37(51-21)57-40-23(3)53-36(18-32(40)46)56-39-22(2)52-35(17-31(39)45)55-27-9-12-41(5)26(16-27)7-8-30-29(41)10-13-42(6)28(11-14-43(30,42)49)25-15-34(47)50-20-25/h15,21-23,26-33,35-40,45-46,48-49H,7-14,16-20H2,1-6H3/t21-,22-,23-,26-,27+,28-,29+,30-,31+,32+,33+,35+,36+,37+,38-,39-,40-,41+,42-,43+/m1/s1
Formel vægt 806.4453