Få mere at vide
Diflunisal Methyl Ester, Mikromol™
Artikelnummer. 30458666
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30458666 4 x 25 mg
30049642 100 mg
30049954 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30458666 Leverandør Mikromol Leverandørnr. MM3050.0100254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Diflunisal Methyl Ester, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Diflunisal Methyl Ester
CAS 55544-00-8
Analyt- eller komponentnavne Diflunisal Methyl Ester
Forsendelsestilstand Room Temperature
Molekylær formel C14 H10 F2 O3
Certificeringer/compliance ISO 17025
Synonym Diflunisal Methyl Ester, 2',4'-Difluoro-4-hydroxy-[1,1'-biphenyl]-3-carboxylic acid methyl ester, Diflunisal methyl ester, Methyl 2',4'-difluoro-4-hydroxybiphenyl-3-carboxylate, Methyl 5-(2,4-difluorophenyl)salicylate
InChI formel InChI=1S/C14H10F2O3/c1-19-14(18)11-6-8(2-5-13(11)17)10-4-3-9(15)7-12(10)16/h2-7,17H,1H3
Løsningstype Neat
SMIL COC(=O)c1cc(ccc1O)c2ccc(F)cc2F
IUPAC navn methyl 5-(2,4-difluorophenyl)-2-hydroxybenzoate
Molekylvægt (g/mol) 264.22
Mængde 4 x 25 mg
Formel vægt 264.0598 g/mol