Få mere at vide
Diflufenican-d3, TRC
Artikelnummer. 30376667
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30376667 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30376667 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4456425MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Diflufenican D3 (3-trifluoromethylphenoxy-2,4,6 D3)
CAS 1185009-29-3
Molekylær formel C19 2H3 H8 F5 N2 O2
Mængde 5 mg
Synonym Diflufenican D3 (3-trifluoromethylphenoxy-2,4,6 D3)
InChI formel InChI=1S/C19H11F5N2O2/c20-12-6-7-16(15(21)10-12)26-17(27)14-5-2-8-25-18(14)28-13-4-1-3-11(9-13)19(22,23)24/h1-10H,(H,26,27)/i3D,4D,9D
SMIL [2H]c1cc([2H])c(c([2H])c1Oc2ncccc2C(=O)Nc3ccc(F)cc3F)C(F)(F)F
IUPAC navn N-(2,4-difluorophenyl)-2-[2,4,6-trideuterio-3-(trifluoromethyl)phenoxy]pyridine-3-carboxamide
Molekylvægt (g/mol) 397.31
Formel vægt 397.0929