missing translation for 'onlineSavingsMsg'
Få mere at vide

Diflucortolone Valerate, LoGiCal

Artikelnummer. 30457527
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30457527 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Artikelnummer. 30457527 Leverandør LoGiCal Leverandørnr. LGCFOR0600.00

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Diflucortolone Valerate
CAS 59198-70-8
Analyt- eller komponentnavne Diflucortolone Valerate
Forsendelsestilstand Room Temperature
Molekylær formel C27 H36 F2 O5
Certificeringer/compliance ISO 9001
Synonym Diflucortolone Valerate, Diflucortolone 21-valerate, Diflucortolone valerate, Neriforte, Nerisona, Nerisone, Nerisone Forte, Temetex, Texmeten, Travazol, (6alpha,11beta,16alpha)-6,9-Difluoro-11-hydroxy-16-methyl-21-[(1-oxopentyl)oxy]pregna-1,4-diene-3,20-dione
InChI formel InChI=1S/C27H36F2O5/c1-5-6-7-23(33)34-14-21(31)24-15(2)10-17-18-12-20(28)19-11-16(30)8-9-26(19,4)27(18,29)22(32)13-25(17,24)3/h8-9,11,15,17-18,20,22,24,32H,5-7,10,12-14H2,1-4H3/t15-,17+,18+,20+,22+,24-,25+,26+,27+/m1/s1
Løsningstype Neat
SMIL CCCCC(=O)OCC(=O)[C@H]1[C@H](C)C[C@H]2[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C
IUPAC navn [2-[(6S,8S,9R,10S,11S,13S,14S,16R,17S)-6,9-difluoro-11-hydroxy-10,13,16-trimethyl-3-oxo-7,8,11,12,14,15,16,17-octahydro-6H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] pentanoate
Industry Type Clinical Testing
Molekylvægt (g/mol) 478.57
Mængde 10 mg
Formel vægt 478.2531 g/mol
Vis mere Vis mindre
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.