Få mere at vide
Difloxacin-d3 (methyl-d3), CDN
Artikelnummer. 30322770
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30322770 0.005 g
30313870 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322770 Leverandør CDN Leverandørnr. CDND79950.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Difloxacin-d3 (methyl-d3)
CAS 1173147-93-7
Molekylær formel C21 D3 H16 F2 N3 O3
Mængde 0.005 g
Synonym 6-Fluoro-1-(4-fluorophenyl)-4-oxo-7-[4-(trideuteriomethyl)piperazin-1-yl]quinoline-3-carboxylic acid, 6-Fluoro-1-(4-fluorophenyl)-4-oxo-7-[4-(methyl-d3)piperazin-1-yl]quinoline-3-carboxylic acid, Difloxacin-D3 (N-methyl-D3), Difloxacin-d3
InChI formel InChI=1 S/C21H19F2N3O3/c1-24-6-8-25(9-7-24)19-11-18-15(10-17(19)23)20(27)16(21(28)29)12-26(18)14-4-2-13(22)3-5-14/h2-5,10-12 H,6-9H2,1H3,(H,28,29)/i1D3
SMIL [2 H]C([2 H])([2 H])N1CCN(CC1)c2cc3N(C=C(C(=O)O)C(=O)c3cc2F)c4ccc(F)cc4
IUPAC navn 6-fluoro-1-(4-fluorophenyl)-4-oxo-7-[4-(trideuteriomethyl)piperazin-1-yl]quinoline-3-carboxylic acid
Molekylvægt (g/mol) 402.4092
Formel vægt 402.1583 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity