Få mere at vide
Difloxacin-d3 Hydrochloride Salt, TRC
Artikelnummer. 17968689
Skift visning
Mængde:
2.5 mg
10 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
17968689 10 mg -
17919309 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 17968689 Leverandør Toronto Research Chemicals Leverandørnr. TRCD44560110MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1173021-89-0
Molekylær formel C21 2H3 H16 F2 N3 O3 . Cl H
Molekylvægt (g/mol) 438.87
Synonym 6-Fluoro-1-(4-fluorophenyl)-4-oxo-7-[4-(trideuteriomethyl)piperazin-1-yl]quinoline-3-carboxylic acid hydrochloride
IUPAC navn 6-fluoro-1-(4-fluorophenyl)-4-oxo-7-[4-(trideuteriomethyl)piperazin-1-yl]quinoline-3-carboxylic acid;hydrochloride
SMIL Cl.[2H]C([2H])([2H])N1CCN(CC1)c2cc3N(C=C(C(=O)O)C(=O)c3cc2F)c4ccc(F)cc4
Tekniske data
Kemisk navn eller materiale Difloxacin hydrochloride D3 (methyl-D3)
Mængde 10 mg
InChI formel InChI=1S/C21H19F2N3O3.ClH/c1-24-6-8-25(9-7-24)19-11-18-15(10-17(19)23)20(27)16(21(28)29)12-26(18)14-4-2-13(22)3-5-14;/h2-5,10-12H,6-9H2,1H3,(H,28,29);1H/i1D3;
Formel vægt 438.135