Få mere at vide
Difenoconazole-d6, TRC
Artikelnummer. 30378223
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30378223 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30378223 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4453725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Difenoconazole-d6
CAS 2748435-80-3
Renhedsgrad noter HPLC
Molekylær formel C19H11D6Cl2N3O3
Mængde 5 mg
Synonym Difenoconazole D6 (1,1,2,3,3,3-propyl D6), Difenoconazole-d6, Difenoconazole-d6 (1,3-dioxolan-4,5,5-d3-4-methyl-d3) (mixture of diastereomers), Difenoconazole-D6 (4-(methyl-D3)-1,3-dioxolan-2-yl-4,5,5-D3), 1-[[2-[2-Chloro-4-(4-chlorophenoxy)phenyl]-4-(methyl-d3)-1,3-dioxolan-2-yl-4,5,5-d3]methyl]-1H-1,2,4-triazole, Dividend-d6, Dividend (fungicide)-d6, Dragon-d6, Plover-d6, Score-d6, Score 10WG-d6, Score EC 250-d6, Sico-d6, Bardos Neu-d6, CGA 169374-d6, 1-[[2-[2-Chloro-4-(4-chlorophenoxy)phenyl]-4-methyl-1,3-dioxolan-2-yl]methyl]-1H-1,2,4-triazole-d6, Difenoconazole D6 (1,1,2,3,3,3-propyl-D6)
InChI formel InChI=1S/C19H17Cl2N3O3/c1-13-9-25-19(27-13,10-24-12-22-11-23-24)17-7-6-16(8-18(17)21)26-15-4-2-14(20)3-5-15/h2-8,11-13H,9-10H2,1H3/i1D3,9D2,13D
SMIL [2H]C([2H])([2H])C1([2H])OC(Cn2cncn2)(OC1([2H])[2H])c3ccc(Oc4ccc(Cl)cc4)cc3Cl
IUPAC navn 1-[[2-[2-chloro-4-(4-chlorophenoxy)phenyl]-4,4,5-trideuterio-5-(trideuteriomethyl)-1,3-dioxolan-2-yl]methyl]-1,2,4-triazole
Molekylvægt (g/mol) 412.3
Formel vægt 411.1
Procent renhed >95