Learn More
BD Difco™ Dehydrated Culture Media: Noble Agar
Product Code. 11798223
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
Product Code. Quantity Unit Size
11798223 500 g 500g
1 options
Product Code. 11798223 Supplier BD Supplier No. 214230
  • ONLINE EXCLUSIVE PRICE
    6625.00 DKK
    4239.49 DKK / 500g
    Spare 2385.51 DKK
    36% Slukket
 Restricted Item
Forventet forsendelse: 22-12-2026
Used as a solidifying agent essentially free of impurities. Difco™ Dehydrated Culture Media: Noble Agar is perfect for electrophoretic and nutritional procedures wherever increased purity is required.
Used as a solidifying agent essentially free of impurities. Difco™ Dehydrated Culture Media: Noble Agar is perfect for electrophoretic and nutritional procedures wherever increased purity is required.
  • Solidifying agent
  • Free of impurities
Applications:
  • Used for electrophoretic and nutritional procedures
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO/DIS, 11014
Concentration 1.5%
Content And Storage <30°C
Additive Noble Agar
Form Dehydrated
For Use With (Application) Preparing microbiological culture media, for Electrophoretic and nutritional procedures
Packaging Type Bottle
Quantity 500 g