Få mere at vide
Diethylstilbestrol Dipropionate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Diethylstilbestrol Dipropionate |
| CAS | 130-80-3 |
| Molekylær formel | C24H28O4 |
| Mængde | 250 mg |
| Synonym | Clinestrol, Cyren B, Dibestil, Diethylstilbestrol Dipropionate, Diethylstilbestrol Propionate, Dihydroxydiethylstilbene Dipropionate, Estilben, Estilbin, Estroben DF, Estrobene DP, Estrogenin, Estrostilben, Euvestin, Gynolett, Horfemine, Neo-Oestranol II, New-Oestranol 11, Oestrogynaedron, Orestol, Pabestrol D, Sinciclan, Sinestrol, Stilbestrol dipropionate, Stilbestrol propionate, Stilbestronate, Stilboestrol DP, Stilbofax, Stilronate, Synestrol, Synoestrol, Synoestron, Syntestrin, Syntestrine, Tauripar B, Vetoestrol, Willestrol, p,p'-Dipropionoxy-trans-alpha,beta-diethylstilbene, alpha,alpha'-Diethyl-4,4'-stilbenediol Dipropionate, alpha,alpha'-Diethyl-4,4'-stilbenediol Dipropionyl Ester |
| InChI formel | InChI=1S/C24H28O4/c1-5-21(17-9-13-19(14-10-17)27-23(25)7-3)22(6-2)18-11-15-20(16-12-18)28-24(26)8-4/h9-16H,5-8H2,1-4H3/b22-21+ |
| SMIL | CCC(=O)Oc1ccc(cc1)\C(=C(/CC)\c2ccc(OC(=O)CC)cc2)\CC |
| Molekylvægt (g/mol) | 380.48 |
| Formel vægt | 380.1988 |