Få mere at vide
Diethylenetriaminepenta(methylenephosphonic acid), technical grade 50%, TRC
Artikelnummer. 30180172
Skift visning
Mængde:
10 mL
25 mL
50 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30180172 25 mL
30180281 50 mL
30180098 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30180172 Leverandør Toronto Research Chemicals Leverandørnr. TRCD44635825ML
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 15827-60-8
Molekylær formel C9H28N3O15P5
Molekylvægt (g/mol) 573.2
Synonym Phosphonic acid, [[(phosphonomethyl)imino]bis[2,1-ethanediylnitrilobis(methylene)]]tetrakis- (9CI), Phosphonic acid, [[bis[2-[bis(phosphonomethyl)amino]ethyl]amino]methyl]- (8CI), P,P’,P’’,P’’’-[[(Phosphonomethyl)imino]bis[2,1-ethanediylnitrilobis(methylene)]]tetrakis[phosphonic acid], CIX, Cublen D 50, Cublen DNC 450, DETPMP, DQ 2060, DTPA-P, DTPF, DTPMP, DTPMPA, DTPP, DTPPA, Dequest 2060, Dequest 2060A, Dequest 2060S, Dequest SPE 9505, Diethylenetriamine-N,N,N',N' ',N' '-penta(methylenephosphonic acid), Diethylenetriamine-N,N,N',N' ',N' '-pentakis(methylenephosphonic acid), Diethylenetriaminepenta(methylenephosphonic acid), Diethylenetriaminepentakis(methylenephosphonic acid), Diethylenetriaminepentakis(methylphosphonic acid), Diethylenetriaminopenta(methylenephosphonic acid), Ethylenetriaminepenta(methylenephosphonic acid), Lonza 905, Sequion 40H50, Sirrix Antox, Versenate PS
IUPAC navn [bis[2-[bis(phosphonomethyl)amino]ethyl]amino]methylphosphonic acid
SMIL OP(=O)(O)CN(CCN(CP(=O)(O)O)CP(=O)(O)O)CCN(CP(=O)(O)O)CP(=O)(O)O
Tekniske data
Kemisk navn eller materiale Diethylenetriaminepenta(methylenephosphonic acid) (technical grade)
Mængde 25 mL
InChI formel InChI=1S/C9H28N3O15P5/c13-28(14,15)5-10(1-3-11(6-29(16,17)18)7-30(19,20)21)2-4-12(8-31(22,23)24)9-32(25,26)27/h1-9H2,(H2,13,14,15)(H2,16,17,18)(H2,19,20,21)(H2,22,23,24)(H2,25,26,27)
Formel vægt 573.0208