Få mere at vide
Diethyleneglycol Monoethyl-d5 Ether, TRC
Artikelnummer. 30369296
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30369296 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30369296 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4443565MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Diethyleneglycol Monoethyl-d5 Ether
CAS 1219804-11-1
Molekylær formel C6 D5 H9 O3
Mængde 5 mg
Synonym Diethyleneglycol Monoethyl-d5 Ether, Diethyleneglycol Monoethyl Ether-d5, Diethyleneglycol Monoethyl Ether-D5 (ethoxy-D5), 2-[2-(Ethoxy-d5)ethoxy]ethanol, 3,6-Dioxa-1-octanol-7,7,8,8,8-d5
InChI formel InChI=1S/C6H14O3/c1-2-8-5-6-9-4-3-7/h7H,2-6H2,1H3/i1D3,2D2
SMIL [2H]C([2H])([2H])C([2H])([2H])OCCOCCO
IUPAC navn 2-[2-(1,1,2,2,2-pentadeuterioethoxy)ethoxy]ethanol
Molekylvægt (g/mol) 139.204
Formel vægt 139.126