Learn More
Diethylcyanamide, 99+%, Thermo Scientific Chemicals
Catalog Number 10620395
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
617-83-4
C5H10N2
98.15
MFCD00001769
ZZTSQZQUWBFTAT-UHFFFAOYSA-N
n,n-diethylcyanamide, cyanamide, diethyl, n-cyanodiethylamine, diethylkyanamid, usaf ek-6326, diethylkyanamid czech, cyanodiethylamine, diethylamino carbonitrile, diethyl-cyanamide, aminodiethylcarbonitrile
12050
diethylcyanamide
CCN(CC)C#N
617-83-4
C5H10N2
98.15
MFCD00001769
ZZTSQZQUWBFTAT-UHFFFAOYSA-N
n,n-diethylcyanamide, cyanamide, diethyl, n-cyanodiethylamine, diethylkyanamid, usaf ek-6326, diethylkyanamid czech, cyanodiethylamine, diethylamino carbonitrile, diethyl-cyanamide, aminodiethylcarbonitrile
12050
diethylcyanamide
CCN(CC)C#N
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Color White
Density 0.8460g/mL
Boiling Point 186°C to 188°C
Flash Point 69°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula (C2H5)2NCN
Refractive Index 1.4210 to 1.423
Quantity 50 g
Beilstein 04, 121
Specific Gravity 0.846
Formula Weight 98.15
Percent Purity 99+%
Physical Form Fine Crystalline Powder
Chemical Name or Material Diethylcyanamide
Show More Show Less
compliance-icons
Product Identifier
  • Diethylcyanamide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.