missing translation for 'onlineSavingsMsg'
Få mere at vide

Diethylammonium Salicylate, LoGiCal

Artikelnummer. 30458360
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30458360 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30458360 Leverandør LoGiCal Leverandørnr. LGCFOR3461.00

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Diethylammonium Salicylate
CAS 4419-92-5
Analyt- eller komponentnavne Diethylammonium Salicylate
Forsendelsestilstand Ice pack (-20°C)
Molekylær formel C7 H6 O3 . C4 H11 N
Certificeringer/compliance ISO 9001
Synonym Benzoic acid, 2-hydroxy-, compd. with N-ethylethanamine (1:1), Salicylic acid, Et2NH salt (6CI), Salicylic acid, compd. with Et2NH (7CI), Salicylic acid, compd. with diethylamine (1:1) (8CI), Diethylamine, compd. with salicylic acid (1:1) (8CI), Ethanamine, N-ethyl-, 2-hydroxybenzoate (9CI), Artrogota, Damsal, Diethylamine salicylate, Diethylammonium salicylate, Salicylate diethylamine, Salicylic acid diethylamine salt
InChI formel InChI=1S/C7H6O3.C4H11N/c8-6-4-2-1-3-5(6)7(9)10;1-3-5-4-2/h1-4,8H,(H,9,10);5H,3-4H2,1-2H3
Løsningstype Neat
SMIL CCNCC.OC(=O)c1ccccc1O
IUPAC navn N-ethylethanamine;2-hydroxybenzoic acid
Industry Type Clinical Testing
Molekylvægt (g/mol) 211.26
Mængde 10 mg
Formel vægt 211.1208 g/mol
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.