missing translation for 'onlineSavingsMsg'
Learn More
Diethylamino hydroxybenzoyl hexyl benzoate, MedChemExpress Product Code.: 16491586

Diethylamino hydroxybenzoyl hexyl benzoate, MedChemExpress

Product Code. p-200096659
500 mg, 500mg
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
This item is not returnable. View return policy

Product Code. 16491586

Brand: Medchem Express HY109656

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Diethylamino hydroxybenzoyl hexyl benzoate is a photostable UV-A absorber.

Diethylamino hydroxybenzoyl hexyl benzoate is a photostable UV-A absorber.

Specifications

Chemical Name or Material Diethylamino hydroxybenzoyl hexyl benzoate
CAS 302776-68-7
Color White
Purity Grade Notes Research
Molecular Formula C24H31NO4
Quantity 500 mg
Synonym DHHB
Solubility Information DMSO : ≥ 50 mg/mL (125.78 mM)
SMILES O=C(OCCCCCC)C1=CC=CC=C1C(C2=CC=C(N(CC)CC)C=C2O)=O
Molecular Weight (g/mol) 397.51
Formula Weight 397.51
Percent Purity 99.88%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Diethylamino hydroxybenzoyl hexyl benzoate, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.