Få mere at vide
Diethylamine-d11, CDN
Artikelnummer. 30312736
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30312736 0.25 g -
30310407 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30312736 Leverandør CDN Leverandørnr. CDND70880.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Diethylamine-D11
CAS 1173019-51-6
Molekylær formel C4 D11 N
Mængde 0.25 g
Synonym Ethan-1,1,2,2,2-d5-amine-d, N-(ethyl-1,1,2,2,2-d5)- (ACI), N-(Ethyl-1,1,2,2,2-d5)ethan-1,1,2,2,2-d5-amine-d (ACI), N,1,1,2,2,2-Hexadeuterio-N-(1,1,2,2,2-pentadeuterioethyl)ethanamine, N-(Ethyl-d5)ethan-d5-amine-d, Diethyl-d10-amine-d, Diethylamine-d11, Diethylamine-D11
InChI formel InChI=1 S/C4H11N/c1-3-5-4-2/h5H,3-4H2,1-2H3/i1D3,2D3,3D2,4D2/hD
SMIL [2 H]N(C([2 H])([2 H])C([2 H])([2 H])[2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H]
IUPAC navn N,1,1,2,2,2-hexadeuterio-N-(1,1,2,2,2-pentadeuterioethyl)ethanamine
Molekylvægt (g/mol) 84.2046
Formel vægt 84.1582 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity