Få mere at vide
Diethyl Phthalate-3,4,5,6-d4, CDN
Artikelnummer. 30320899
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30320899 0.1 g 0.1g
30318575 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320899 Leverandør CDN Leverandørnr. CDND27260.1G
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Phthalic acid, bis-ethyl ester D4
CAS 93952-12-6
Molekylær formel C12 2H4 H10 O4
Mængde 0.1 g
Synonym 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid, diethyl ester (9 CI), Phthalic acid diethyl ester (3,4,5,6)-D4, Phthalic acid, bis-ethyl ester D4, Diethyl phthalate-d4
InChI formel InChI=1 S/C12H14O4/c1-3-15-11(13)9-7-5-6-8-10(9)12(14)16-4-2/h5-8 H,3-4H2,1-2H3/i5D,6 D,7 D,8 D
SMIL [2 H]c1c([2 H])c([2 H])c(C(=O)OCC)c(C(=O)OCC)c1[2 H]
IUPAC navn diethyl 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate
Molekylvægt (g/mol) 226.26
Formel vægt 226.1143 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity