Få mere at vide
Diethyl Phosphate Sodium Salt, TRC
Artikelnummer. 30134103
Skift visning
Mængde:
250 mg
2.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134103 2.5 g
30005321 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30134103 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4447202.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2870-30-6
Molekylær formel C4H10NaO4P
Molekylvægt (g/mol) 176.08
Synonym Phosphoric Acid Diethyl Ester Sodium Salt, Ethyl Sodium Phosphate, Diethyl Sodium Phosphate, Sodium Diethyl Phosphate;
SMIL CCOP(=O)([O-])OCC.[Na+]
Tekniske data
Kemisk navn eller materiale Diethyl Phosphate Sodium Salt
Renhedsgrad noter HPLC
Mængde 2.5 g
InChI formel InChI=1S/C4H11O4P.Na/c1-3-7-9(5,6)8-4-2;/h3-4H2,1-2H3,(H,5,6);/q;+1/p-1
Formel vægt 176.0214
Procent renhed >95%