Få mere at vide
Diethyl Methylphosphonate-13C, TRC
Artikelnummer. 30379787
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30379787 10 mg
1 options
Artikelnummer. 30379787 Leverandør Toronto Research Chemicals Leverandørnr. TRCD44235710MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Diethyl Methylphosphonate-13C
CAS 1342310-32-0
Molekylær formel C413CH13O3P
Mængde 10 mg
Synonym Bis(diethoxyphosphinyl)methane-13C, DEMP-13C, Diethoxymethylphosphine Oxide-13C, Diethyl Methanephosphonate-13C, O,O-Diethyl Methylphosphonate-13C, P-Methyl-phosphonic Acid Diethyl Ester-13C
InChI formel InChI=1S/C5H13O3P/c1-4-7-9(3,6)8-5-2/h4-5H2,1-3H3/i3+1
SMIL [13CH3]P(=O)(OCC)OCC
Molekylvægt (g/mol) 153.12