Få mere at vide
Diethyl Malonate-2-13C, TRC
Artikelnummer. 30134642
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134642 50 mg
30133354 100 mg
30009399 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30134642 Leverandør Toronto Research Chemicals Leverandørnr. TRCD44442150MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 67035-94-3
Molekylær formel C613CH12O4
Molekylvægt (g/mol) 161.16
Synonym Diethyl Ester Propanedioic-2-13C Acid, Propanedioic Acid 1,3-Diethyl Ester-13C, Malonic Acid Diethyl Ester-13C, Propanedioic Acid Diethyl Ester-13C, Carbethoxyacetic Ester, Dicarbethoxymethane-13C, Diethyl Propane-1,3-dioate-13C, Diethyl Propanedioate-13C;Propanedioic-1,3-13C2 Acid Diethyl Ester
SMIL O=C(OCC)[13CH2]C(OCC)=O
Tekniske data
Kemisk navn eller materiale Diethyl Malonate-2-13C
Mængde 50 mg
InChI formel InChI=1S/C7H12O4/c1-3-10-6(8)5-7(9)11-4-2/h3-5H2,1-2H3/i5+1