Få mere at vide
Diethyl (Ethoxycarbonyl)phosphonate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Ethyl diethoxyphosphinylformate |
| CAS | 1474-78-8 |
| Molekylær formel | C7 H15 O5 P |
| Mængde | 5 g |
| Synonym | Phosphinecarboxylic acid, 1,1-diethoxy-, ethyl ester, 1-oxide, Formic acid, phosphono-, triethyl ester (7CI,8CI), Phosphinecarboxylic acid, diethoxy-, ethyl ester, oxide (9CI), Diethyl (ethoxycarbonyl)phosphonate, Diethyl carbethoxyphosphonate, Ethyl diethylphosphonoformate, NSC 108684, Triethyl carboxyphosphonate, Triethyl phosphonoformate, Ethyl (Diethoxyphosphoryl)formate, O,O-Diethyl ethoxycarbonylphosphonate |
| InChI formel | InChI=1S/C7H15O5P/c1-4-10-7(8)13(9,11-5-2)12-6-3/h4-6H2,1-3H3 |
| SMIL | CCOC(=O)P(=O)(OCC)OCC |
| IUPAC navn | ethyl diethoxyphosphorylformate |
| Molekylvægt (g/mol) | 210.16 |
| Formel vægt | 210.0657 |