Få mere at vide
Diethyl Ether, Dried, Reagent Grade, BHT Stabilized, Scharlab
Artikelnummer. 11314084
Skift visning
Mængde:
1 L
Emballage:
Glass Bottle
Pakningsstørrelse:
1 L
Artikelnummer. Mængde Emballage unitSize
11314084 1 L Glass Bottle 1 L
1 options
Artikelnummer. 11314084 Leverandør Scharlab Leverandørnr. ET00801000
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Contains a maximum of 0.0075% H2O and stabilized with approximately 7 ppm of 2,6-Di-tert-butyl-4-methylphenol.
Reagent grade solvents are the most frequently used grade for laboratory analytical work. They correspond to a grade of high quality chemicals for laboratory and specialized industrial use.
Tekniske data
Kemisk navn eller materiale Diethyl ether
Smeltepunkt -116.3°C
CAS 60-29-7
Farve Colorless
Surhed Max. 0.0002meq/g
Alkalinitet Max. 0.0002meq/g
Udseende Clear
Tæthed 0.713 - 0.715g/cm3
Mængde 1 L
Flammepunkt −40°C
Identifikation Passes test
Tændingspunkt 170°C
Emballage Glass Bottle
Fysisk form Liquid
Renhedsgrad noter Applications: solvents, synthesis of organic products, solvent for active principles from plant and animal tissues extractions
Rester efter fordampning Max. 0.001%
Molekylær formel C4H10O
Damptryk 587 hPa
MDL nummer MFCD00011646
FN nummer 1155
Vand Max. 0.0075%, Dried
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
Kogepunkt 34.6°C
Opløselighedsinformation 69g/L in water
InChI nøgle RTZKZFJDLAIYFH-UHFFFAOYSA-N
SMIL CCOCC
IUPAC navn ethoxyethane
Molekylvægt (g/mol) 74.12
PubChem CID 3283
ChEBI CHEBI:35702
Formel vægt 74.12g/mol
Procent renhed Min. 99.5%
Grad Reagent