Få mere at vide
Diethyl-d10 Chlorophosphate, CDN
Artikelnummer. 30312369
Skift visning
Mængde:
0.01 g
Artikelnummer. Mængde
30312369 0.01 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30312369 Leverandør CDN Leverandørnr. CDND80810.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Diethyl Chlorophosphate-D10
CAS 141870-47-5
Molekylær formel C4 D10 Cl O3 P
Mængde 0.01 g
Synonym Phosphorochloridic acid, di(ethyl-d5) ester (9 CI), Phosphorochloridic acid di(ethyl-d5) ester, Diethyl Chlorophosphate-D10, Diethyl-d10 Chlorophosphate
InChI formel InChI=1 S/C4H10ClO3P/c1-3-7-9(5,6)8-4-2/h3-4H2,1-2H3/i1D3,2D3,3D2,4D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])OP(=O)(Cl)OC([2 H])([2 H])C([2 H])([2 H])[2 H]
IUPAC navn 1-[chloro(1,1,2,2,2-pentadeuterioethoxy)phosphoryl]oxy-1,1,2,2,2-pentadeuterioethane
Molekylvægt (g/mol) 182.6088
Formel vægt 182.0684 g/mol
Procent renhed 99 atom % D, min 97% Chemical Purity