Få mere at vide
Diethyl-d10-amine HBr, CDN
Artikelnummer. 30316104
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30316104 1 g -
30312821 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30316104 Leverandør CDN Leverandørnr. CDND71381G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Diethylamine-D10 Hydrobromide
CAS 1219803-53-8
Molekylær formel C4 D10 H N . Br H
Mængde 1 g
Synonym 1,1,2,2,2-Pentadeuterio-N-(1,1,2,2,2-pentadeuterioethyl)ethanamine hydrobromide, N-(Ethyl-d5)ethan-d5-amine hydrobromide, Diethyl-d10-amine hydrobromide, Diethylamine-d10 hydrobromide, Diethylamine-D10 hydrobromide
InChI formel InChI=1 S/C4H11N.BrH/c1-3-5-4-2;/h5H,3-4H2,1-2H3;1 H/i1D3,2D3,3D2,4D2;
SMIL Br.[2 H]C([2 H])([2 H])C([2 H])([2 H])NC([2 H])([2 H])C([2 H])([2 H])[2 H]
IUPAC navn 1,1,2,2,2-pentadeuterio-N-(1,1,2,2,2-pentadeuterioethyl)ethanamine;hydrobromide
Molekylvægt (g/mol) 164.1104
Formel vægt 163.0781 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity