Learn More
Diethyl Carbonate, Sigma-Aldrich™
Product Code. 15868870
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
Unit Size:
1 L
100mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15868870 1 L 1 L
15848870 100 mL 100mL
15858870 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15868870 Supplier Sigma Aldrich Supplier No. 8028981000
 Restricted Item
Forventet forsendelse: 23-11-2026
For synthesis
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is used as a greener reagent.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 105-58-8
Specifications
Melting Point -43°C
Density 0.97 g/cm3 at 20°C
Boiling Point 126°C/1013 hPa
Molecular Formula CO(OC2H5)2
MDL Number MFCD00009107
Quantity 1 L
Synonym Diethyl carbonate, Carbonic acid diethyl ester, Diethyl carbonate
Molecular Weight (g/mol) 118.13
Grade Research