missing translation for 'onlineSavingsMsg'
Få mere at vide

Diethyl (Boc-amino)malonat, TRC

Artikelnummer. 30376477
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30376477 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30376477 Supplier Toronto Research Chemicals Supplier No. TRCD443800500MG
 Begrænset vare
Forventet forsendelse: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Højrenheds organiske molekyler og analytiske standarder, strategisk leveret verden over for at muliggøre innovation og kommerciel succes.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Diethyl (Boc-amino)malonate
CAS 102831-44-7
Molekylær formel C12 H21 N O6
Mængde 500 mg
InChI formel InChI=1S/C12H21NO6/c1-6-17-9(14)8(10(15)18-7-2)13-11(16)19-12(3,4)5/h8H,6-7H2,1-5H3,(H,13,16)
SMIL CCOC(=O)C(NC(=O)OC(C)(C)C)C(=O)OCC
IUPAC navn diethyl 2-[(2-methylpropan-2-yl)oxycarbonylamino]propanedioat
Molekylvægt (g/mol) 275.3
Formel vægt 275.1369
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.