missing translation for 'onlineSavingsMsg'
Learn More

Diethyl benzylmalonate, 97%, Thermo Scientific Chemicals

Catalog Number p-7026720
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
607-81-8
C14H18O4
250.294
MFCD00009166
ICZLTZWATFXDLP-UHFFFAOYSA-N
diethyl benzylmalonate, diethyl 2-benzylmalonate, benzylmalonic acid diethyl ester, propanedioic acid, phenylmethyl-, diethyl ester, unii-40y0x0xn6p, malonic acid, benzyl-, diethyl ester, malonic acid, 2-benzyl-, diethyl ester, diethyl benzyl malonate, 1,3-diethyl 2-benzylpropanedioate, diethyl benzylpropanedioate
69090
diethyl 2-benzylpropanedioate
CCOC(=O)C(CC1=CC=CC=C1)C(=O)OCC
This item is not returnable. View return policy
607-81-8
C14H18O4
250.294
MFCD00009166
ICZLTZWATFXDLP-UHFFFAOYSA-N
diethyl benzylmalonate, diethyl 2-benzylmalonate, benzylmalonic acid diethyl ester, propanedioic acid, phenylmethyl-, diethyl ester, unii-40y0x0xn6p, malonic acid, benzyl-, diethyl ester, malonic acid, 2-benzyl-, diethyl ester, diethyl benzyl malonate, 1,3-diethyl 2-benzylpropanedioate, diethyl benzylpropanedioate
69090
diethyl 2-benzylpropanedioate
CCOC(=O)C(CC1=CC=CC=C1)C(=O)OCC

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.069
Boiling Point 162°C to 163°C (10 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.486
Quantity 25 g
Beilstein 615793
Formula Weight 250.29
Percent Purity 97%
Chemical Name or Material Diethyl benzylmalonate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'