missing translation for 'onlineSavingsMsg'
Learn More

Diethyl allyl(3-phenyl-2-propynyl)malonate, 96%, Thermo Scientific Chemicals

Catalog Number p-7030156
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
143633-91-4
C19H22O4
314.381
MFCD09953493
ZSPBIVGRCZUELN-UHFFFAOYSA-N
diethyl allyl 3-phenyl-2-propynyl malonate, diethyl 2-allyl-2-3-phenylprop-2-yn-1-yl malonate, propanedioic acid,2-3-phenyl-2-propyn-1-yl-2-2-propen-1-yl-, 1,3-diethyl ester, 1,3-diethyl 2-3-phenylprop-2-yn-1-yl-2-prop-2-en-1-yl propanedioate, acmc-20ed36, 3-phenylpropargylallylmalonic acid diethyl ester, allyl 3-phenyl-2-propynyl malonic acid diethyl ester, diethyl 1-phenyl-6-heptene-1-yne-4,4-dicarboxylate, diethyl 3-phenylprop-2-yn-1-yl prop-2-en-1-yl propanedioate
10086992
diethyl 2-(3-phenylprop-2-ynyl)-2-prop-2-enylpropanedioate
CCOC(=O)C(CC=C)(CC#CC1=CC=CC=C1)C(=O)OCC
This item is not returnable. View return policy
143633-91-4
C19H22O4
314.381
MFCD09953493
ZSPBIVGRCZUELN-UHFFFAOYSA-N
diethyl allyl 3-phenyl-2-propynyl malonate, diethyl 2-allyl-2-3-phenylprop-2-yn-1-yl malonate, propanedioic acid,2-3-phenyl-2-propyn-1-yl-2-2-propen-1-yl-, 1,3-diethyl ester, 1,3-diethyl 2-3-phenylprop-2-yn-1-yl-2-prop-2-en-1-yl propanedioate, acmc-20ed36, 3-phenylpropargylallylmalonic acid diethyl ester, allyl 3-phenyl-2-propynyl malonic acid diethyl ester, diethyl 1-phenyl-6-heptene-1-yne-4,4-dicarboxylate, diethyl 3-phenylprop-2-yn-1-yl prop-2-en-1-yl propanedioate
10086992
diethyl 2-(3-phenylprop-2-ynyl)-2-prop-2-enylpropanedioate
CCOC(=O)C(CC=C)(CC#CC1=CC=CC=C1)C(=O)OCC

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Refractive Index 1.514
Quantity 1 g
Formula Weight 314.38
Percent Purity 96%
Chemical Name or Material Diethyl allyl(3-phenyl-2-propynyl)malonate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'