Få mere at vide
Diethyl 4-(2,3-Dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate, Mikromol™
Artikelnummer. 30459562
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30459562 4 x 25 mg
30051392 100 mg
30051541 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30459562 Leverandør Mikromol Leverandørnr. MM0139.0300254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Diethyl 4-(2,3-Dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Diethyl 4-(2,3-Dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
CAS 79925-38-5
Analyt- eller komponentnavne Diethyl 4-(2,3-Dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
Forsendelsestilstand Room Temperature
Molekylær formel C19 H21 Cl2 N O4
Certificeringer/compliance ISO 17025
Synonym Felodipine Imp. C (EP), Diethyl 4-(2,3-Dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate, Felodipine Impurity C
InChI formel InChI=1S/C19H21Cl2NO4/c1-5-25-18(23)14-10(3)22-11(4)15(19(24)26-6-2)16(14)12-8-7-9-13(20)17(12)21/h7-9,16,22H,5-6H2,1-4H3
Løsningstype Neat
SMIL CCOC(=O)C1=C(C)NC(=C(C1c2cccc(Cl)c2Cl)C(=O)OCC)C
IUPAC navn diethyl 4-(2,3-dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
Molekylvægt (g/mol) 398.28
Mængde 4 x 25 mg
Formel vægt 397.0848 g/mol