missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 2-thienylmethylphosphonate, 95%, Thermo Scientific Chemicals

Catalog Number p-7056360
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
2026-42-8
C9H15O3PS
234.25
MFCD00128457
GOJBUVQDOFHBLF-UHFFFAOYSA-N
diethyl thiophen-2-ylmethyl phosphonate, diethyl 2-thienylmethyl phosphonate, thiophen-2-ylmethyl-phosphonic acid diethyl ester, diethyl 2-thienylmethylphosphonate, 2-diethoxyphosphorylmethyl thiophene, phosphonic acid, 2-thienylmethyl-, diethyl ester, diethyl thiophen-2-ylmethylphosphonate, ksc207o5j, 2-diethoxyphosphinylmethyl thiophene
4185897
2-(diethoxyphosphorylmethyl)thiophene
CCOP(=O)(CC1=CC=CS1)OCC
This item is not returnable. View return policy
2026-42-8
C9H15O3PS
234.25
MFCD00128457
GOJBUVQDOFHBLF-UHFFFAOYSA-N
diethyl thiophen-2-ylmethyl phosphonate, diethyl 2-thienylmethyl phosphonate, thiophen-2-ylmethyl-phosphonic acid diethyl ester, diethyl 2-thienylmethylphosphonate, 2-diethoxyphosphorylmethyl thiophene, phosphonic acid, 2-thienylmethyl-, diethyl ester, diethyl thiophen-2-ylmethylphosphonate, ksc207o5j, 2-diethoxyphosphinylmethyl thiophene
4185897
2-(diethoxyphosphorylmethyl)thiophene
CCOP(=O)(CC1=CC=CS1)OCC

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow
Refractive Index 1.52
Quantity 250 mg
Beilstein 1683411
Formula Weight 234.25
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Diethyl 2-thienylmethylphosphonate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'