Få mere at vide
Diethyl 2-Methyl-d3-2-propylmalonate, TRC
Artikelnummer. 30369294
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30369294 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30369294 Leverandør Toronto Research Chemicals Leverandørnr. TRCD44450710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Diethyl 2-Methyl-d3-2-propylmalonate
CAS 1246815-67-7
Molekylær formel C11H17D3O4
Mængde 10 mg
Synonym Methylpropyl-malonic Acid Diethyl Ester-d3, Diethyl Methylpropylmalonate-d3
InChI formel InChI=1S/C11H20O4/c1-5-8-11(4,9(12)14-6-2)10(13)15-7-3/h5-8H2,1-4H3/i4D3
SMIL [2H]C([2H])([2H])C(CCC)(C(=O)OCC)C(=O)OCC
IUPAC navn diethyl 2-propyl-2-(trideuteriomethyl)propanedioate
Molekylvægt (g/mol) 219.29
Formel vægt 219.155