missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 1-tetradecylphosphonate, 97+%, Thermo Scientific Chemicals

Catalog Number 11499397
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
5191-09-3
MFCD00015597
CPWIYYOBPHXJIA-UHFFFAOYSA-N
640154
CCCCCCCCCCCCCCP(=O)(OCC)OCC
This item is not returnable. View return policy
5191-09-3
MFCD00015597
CPWIYYOBPHXJIA-UHFFFAOYSA-N
640154
CCCCCCCCCCCCCCP(=O)(OCC)OCC

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 8°C
Density 0.922
Boiling Point 199°C to 201°C (3 mmHg)
Molecular Formula C18H39O3P
Linear Formula CH3(CH2)13P(O)(OCH2CH3)2
Refractive Index 1.4445
Quantity 100 g
Synonym diethyl 1-tetradecanephosphonate, diethyl 1-tetradecylphosphonate, diethyl tetradecylphosphonate, acmc-20al9h, diethyl tetradecane-1-phosphonate, tetradecylphosphonic acid diethyl ester, tetradecyl-phosphonic acid diethyl ester, 1-tetradecanephosphonic acid diethyl ester, phosphonic acid, tetradecyl-, diethyl ester
IUPAC Name 1-diethoxyphosphoryltetradecane
Molecular Weight (g/mol) 334.48
Formula Weight 334.47
Percent Purity 98%
Chemical Name or Material Diethyl 1-tetradecylphosphonate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'