Få mere at vide
Diethyl-1,1,1',1'-d4-stilbestrol-3,3',5,5'-d4, CDN
Artikelnummer. 30320901
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30320901 0.01 g
30311051 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320901 Leverandør CDN Leverandørnr. CDND28490.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale trans-Diethyl-1,1,1',1'-stilbestrol-3,3',5,5'-d8
CAS 91318-10-4
Molekylær formel C18 2H8 H12 O2
Mængde 0.01 g
Synonym Phen-2,6-d2-ol, 4,4'-[(1 E)-1,2-di(ethyl-1,1-d2)-1,2-ethenediyl]bis-, Phen-2,6-d2-ol, 4,4'-[1,2-di(ethyl-1,1-d2)-1,2-ethenediyl]bis-, (E)- (9 CI), 4,4'-[(1 E)-1,2-Di(ethyl-1,1-d2)-1,2-ethenediyl]bis[phen-2,6-d2-ol], trans-Diethyl-1,1,1',1'-stilbestrol-3,3',5,5'-d8, (E)-Diethyl-1,1,1',1'-d4-stilbestrol-3,3',5,5'-d4, Diethylstilbestrol (ring-3,3',5,5'-diethyl-1,1,1',1'-D8), Diethylstilbestrol (ring-3,3',5,5'-diethyl-1,1,1',1'-D8), Diethylstilbestrol (ring-3,3',5,5'-diethyl-1,1,1',1'-D8)
InChI formel InChI=1 S/C18H20O2/c1-3-17(13-5-9-15(19)10-6-13)18(4-2)14-7-11-16(20)12-8-14/h5-12,19-20 H,3-4H2,1-2H3/b18-17+/i3D2,4D2,9 D,10 D,11 D,12 D
SMIL [2 H]c1cc(cc([2 H])c1O)\C(=C(\c2cc([2 H])c(O)c([2 H])c2)/C([2 H])([2 H])C)\C([2 H])([2 H])C
IUPAC navn 2,6-dideuterio-4-[(E)-2,2,5,5-tetradeuterio-4-(3,5-dideuterio-4-hydroxyphenyl)hex-3-en-3-yl]phenol
Molekylvægt (g/mol) 276.4
Formel vægt 276.1965 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity