Få mere at vide
Diethazine Hydrochloride, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Diethazine Hydrochloride |
| CAS | 341-70-8 |
| Analyt- eller komponentnavne | Diethazine Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C18 H22 N2 S . Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | 10H-Phenothiazine-10-ethanamine, N,N-diethyl-, hydrochloride (1:1), 10H-Phenothiazine-10-ethanamine, N,N-diethyl-, monohydrochloride (9CI), Phenothiazine, 10-[2-(diethylamino)ethyl]-, monohydrochloride (8CI), Antipar, Aparkazin, Casantin, Deparkin, Diethazine hydrochloride, Diethazine monohydrochloride, Diethazinium chloride, Dinezin, Dinezine, Diparcol, Latibon, Thiantan, Thiontan |
| InChI formel | InChI=1S/C18H22N2S.ClH/c1-3-19(4-2)13-14-20-15-9-5-7-11-17(15)21-18-12-8-6-10-16(18)20;/h5-12H,3-4,13-14H2,1-2H3;1H |
| Løsningstype | Neat |
| SMIL | Cl.CCN(CC)CCN1c2ccccc2Sc3ccccc13 |
| Vis mere |